Assay Details
Gene Symbol: INTS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636036.1 | XP_023491804.1 | 1734 | 76 | ||
XM_023636037.1 | XP_023491805.1 | 2046 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
