Assay Details
Gene Symbol: PUSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636046.1 | XP_023491814.1 | 1034 | 66 | ||
XM_023636047.1 | XP_023491815.1 | 1031 | 66 | |||
XM_023636048.1 | XP_023491816.1 | 1368 | 66 | |||
XM_023636049.1 | XP_023491817.1 | 973 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
