Assay Details
Gene Symbol: C1QTNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496492.6 | XP_001496542.3 | 652 | 109 | ||
XM_023636044.1 | XP_023491812.1 | 344 | 109 |

More Information
Set Membership: |
![]() ![]() |
