Assay Details
Gene Symbol: TNFRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503562.5 | XP_001503612.3 | 499 | 53 | ||
XM_005607600.3 | XP_005607657.1 | 498 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
