Assay Details
Gene Symbol: PINX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496538.5 | XP_001496588.1 | 607 | 64 | ||
GenBank mRNA | JL636841.1 | - | 1 - 2 | 585 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
