Assay Details
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498986.4 | XP_001499036.1 | 289 | 75 | ||
XM_005607730.3 | XP_005607787.1 | 363 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
