Assay Details
Gene Symbol: CBR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499202.5 | XP_001499252.1 | 455 | 94 | ||
XM_005607736.3 | XP_005607793.1 | 455 | 94 | |||
XM_014737758.2 | XP_014593244.1 | 455 | 94 | |||
XM_023636322.1 | XP_023492090.1 | 455 | 94 | |||
XM_023636323.1 | XP_023492091.1 | 455 | 94 | |||
XM_023636324.1 | XP_023492092.1 | 455 | 94 | |||
XM_023636325.1 | XP_023492093.1 | 455 | 94 | |||
XM_023636326.1 | XP_023492094.1 | 455 | 94 | |||
GenBank mRNA | JL624000.1 | - | 1 - 2 | 131 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
