Assay Details
Gene Symbol: PRMT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915180.5 | XP_001915215.2 | 1542 | 100 | ||
XM_014737892.2 | XP_014593378.2 | 1543 | 100 | |||
XM_023636459.1 | XP_023492227.1 | 1544 | 100 | |||
XM_023636460.1 | XP_023492228.1 | 715 | 100 | |||
XM_023636461.1 | XP_023492229.1 | 1069 | 100 | |||
GenBank mRNA | JL636104.1 | - | 1 - 2 | 142 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
