Assay Details
Gene Symbol: ELMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607831.3 | XP_005607888.1 | 669 | 79 | ||
XM_014737969.2 | XP_014593455.1 | 678 | 79 | |||
XM_014737970.2 | XP_014593456.1 | 714 | 79 | |||
XM_014737971.2 | XP_014593457.1 | 779 | 79 | |||
XM_023636514.1 | XP_023492282.1 | 544 | 79 | |||
XM_023636516.1 | XP_023492284.1 | 642 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
