Assay Details
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488066.3 | XP_001488116.2 | 1472 | 59 | ||
XM_014734696.2 | XP_014590182.1 | 56 | ||||
XM_023624358.1 | XP_023480126.1 | 1274 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
