Assay Details
Gene Symbol: BPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624368.1 | XP_023480136.1 | 726 | 71 | ||
GenBank mRNA | JL616815.1 | - | 2 - 3 | 322 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
