Assay Details
Gene Symbol: RIPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363744.4 | XP_003363792.2 | 912 | 63 | ||
XM_005603606.3 | XP_005603663.2 | 909 | 63 | |||
XM_014734222.2 | XP_014589708.1 | 806 | 63 | |||
XM_023624535.1 | XP_023480303.1 | 852 | 63 | |||
XM_023624536.1 | XP_023480304.1 | 717 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
