Assay Details
Gene Symbol: LOC100067842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624544.1 | XP_023480312.1 | 2029 | 98 | ||
GenBank mRNA | JL628456.1 | - | 2 - 3 | 178 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
