Assay Details
Gene Symbol: AGPAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492068.5 | XP_001492118.2 | 605 | 84 | ||
XM_005603762.3 | XP_005603819.1 | 620 | 84 | |||
XM_005603763.3 | XP_005603820.1 | 2840 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
