Assay Details
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493006.6 | XP_001493056.2 | 229 | 59 | ||
XM_005603790.3 | XP_005603847.1 | 229 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
