Assay Details
Gene Symbol: PPIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500007.4 | XP_001500057.1 | 414 | 97 | ||
GenBank mRNA | JL616261.2 | - | 3 - 4 | 293 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
