Assay Details
Gene Symbol: RNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624866.1 | XP_023480634.1 | 1101 | 75 | ||
XM_023624867.1 | XP_023480635.1 | 981 | 75 | |||
XM_023624868.1 | XP_023480636.1 | 1103 | 75 | |||
XM_023624869.1 | XP_023480637.1 | 981 | 75 | |||
XM_023624870.1 | XP_023480638.1 | 974 | 75 | |||
XM_023624872.1 | XP_023480640.1 | 971 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
