Assay Details
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603910.3 | XP_005603967.1 | 318 | 78 | ||
XM_023624927.1 | XP_023480695.1 | 584 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
