Assay Details
Gene Symbol: ENPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502620.4 | XP_001502670.1 | 1211 | 105 | ||
XM_023625037.1 | XP_023480805.1 | 1241 | 105 | |||
XM_023625039.1 | XP_023480807.1 | 1061 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
