Assay Details
Gene Symbol: CILK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001918274.4 | XP_001918309.1 | 941 | 68 | ||
XM_023625092.1 | XP_023480860.1 | 1036 | 68 | |||
XM_023625093.1 | XP_023480861.1 | 942 | 68 | |||
XM_023625094.1 | XP_023480862.1 | 442 | 68 |

More Information
Set Membership: |
![]() ![]() |
