Assay Details
Gene Symbol: ELOVL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734495.2 | XP_014589981.1 | 952 | 81 | ||
XM_014734499.2 | XP_014589985.1 | 970 | 81 | |||
XM_023625098.1 | XP_023480866.1 | 981 | 81 | |||
XM_023625099.1 | XP_023480867.1 | 1016 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
