Assay Details
Gene Symbol: EPHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001914787.4 | XP_001914822.2 | 945 | 56 | ||
XM_023625327.1 | XP_023481095.1 | 1101 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
