Assay Details
Gene Symbol: PGPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625489.1 | XP_023481257.1 | 1150 | 72 | ||
XM_023625490.1 | XP_023481258.1 | 1198 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
