Assay Details
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625558.1 | XP_023481326.1 | 4324 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
