Assay Details
Gene Symbol: ZNF131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625689.1 | XP_023481457.1 | 1410 | 94 | ||
XM_023625690.1 | XP_023481458.1 | 1414 | 94 | |||
XM_023625691.1 | XP_023481459.1 | 1405 | 94 | |||
XM_023625692.1 | XP_023481460.1 | 1534 | 94 | |||
XM_023625693.1 | XP_023481461.1 | 1494 | 94 | |||
XM_023625694.1 | XP_023481462.1 | 1307 | 94 | |||
XM_023625695.1 | XP_023481463.1 | 1249 | 94 | |||
XM_023625696.1 | XP_023481464.1 | 1179 | 94 | |||
XM_023625697.1 | XP_023481465.1 | 489 | 94 | |||
GenBank mRNA | JL621009.1 | - | 2 - 3 | 346 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
