Assay Details
Gene Symbol: TTC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499030.6 | XP_001499080.1 | 514 | 85 | ||
XM_023625705.1 | XP_023481473.1 | 465 | 85 | |||
GenBank mRNA | JL623926.1 | - | 1 - 2 | 73 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
