Assay Details
Gene Symbol: FYB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604297.3 | XP_005604354.1 | 1948 | 72 | ||
XM_005604298.3 | XP_005604355.1 | 1948 | 72 | |||
GenBank mRNA | JL637332.1 | - | 6 - 7 | 508 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
