Assay Details
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625791.1 | XP_023481559.1 | 215 | 60 | ||
XM_023625792.1 | XP_023481560.1 | 343 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
