Assay Details
Gene Symbol: CDH18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501055.5 | XP_001501105.1 | 1393 | 72 | ||
XM_023625812.1 | XP_023481580.1 | 2093 | 72 | |||
XM_023625813.1 | XP_023481581.1 | 6248 | 72 | |||
XM_023625814.1 | XP_023481582.1 | 1393 | 72 | |||
XM_023625815.1 | XP_023481583.1 | 1393 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
