Assay Details
Gene Symbol: PDCD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734952.2 | XP_014590438.1 | 460 | 55 | ||
XM_023625912.1 | XP_023481680.1 | 568 | 55 | |||
XM_023625913.1 | XP_023481681.1 | 576 | 55 | |||
XM_023625914.1 | XP_023481682.1 | 579 | 55 | |||
XM_023625915.1 | XP_023481683.1 | 579 | 55 | |||
XM_023625916.1 | XP_023481684.1 | 578 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014734952.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
