Assay Details
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502654.6 | XP_001502704.1 | 1529 | 85 | ||
XM_023643272.1 | XP_023499040.1 | 1263 | 85 | |||
XM_023643273.1 | XP_023499041.1 | 1106 | 85 | |||
XM_023643274.1 | XP_023499042.1 | 1170 | 85 | |||
XM_023643275.1 | XP_023499043.1 | 1491 | 85 | |||
XM_023643276.1 | XP_023499044.1 | 1237 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
