Assay Details
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740980.2 | XP_014596466.1 | 355 | 54 | ||
XM_014740982.2 | XP_014596468.1 | 355 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
