Assay Details
Gene Symbol: RHEBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504155.4 | XP_001504205.1 | 588 | 82 | ||
XM_014740814.2 | XP_014596300.2 | 465 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
