Assay Details
Gene Symbol: NAA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604452.3 | XP_005604509.2 | 239 | 79 | ||
XM_023626025.1 | XP_023481793.1 | 487 | 79 | |||
XM_023626026.1 | XP_023481794.1 | 259 | 79 | |||
XM_023626027.1 | XP_023481795.1 | 391 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
