Assay Details
Gene Symbol: POLR3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626062.1 | XP_023481830.1 | 403 | 67 | ||
XM_023626063.1 | XP_023481831.1 | 403 | 67 | |||
XM_023626064.1 | XP_023481832.1 | 381 | 67 | |||
XM_023626065.1 | XP_023481833.1 | 273 | 67 | |||
XM_023626066.1 | XP_023481834.1 | 247 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
