Assay Details
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496329.5 | XP_001496379.2 | 205 | 54 | ||
XM_014735064.2 | XP_014590550.2 | 211 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
