Assay Details
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497470.4 | XP_001497520.1 | 717 | 128 | ||
XM_023626235.1 | XP_023482003.1 | 717 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
