Assay Details
Gene Symbol: FKBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626260.1 | XP_023482028.1 | 431 | 98 | ||
XM_023626261.1 | XP_023482029.1 | 519 | 98 | |||
GenBank mRNA | JL619482.1 | - | 1 - 2 | 178 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
