Assay Details
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499130.6 | XP_001499180.3 | 202 | 88 | ||
XM_005604583.3 | XP_005604640.1 | 211 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005604583.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
