Assay Details
Gene Symbol: PROCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604593.3 | XP_005604650.1 | 819 | 61 | ||
XM_005604594.3 | XP_005604651.1 | 707 | 61 | |||
XM_005604595.3 | XP_005604652.1 | 876 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
