Assay Details
Gene Symbol: SRSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626595.1 | XP_023482363.1 | 730 | 71 | ||
GenBank mRNA | HQ890300.1 | - | 1 - 2 | 76 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
