Assay Details
Gene Symbol: YWHAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626621.1 | XP_023482389.1 | 889 | 72 | ||
XM_023626622.1 | XP_023482390.1 | 705 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
