Assay Details
Gene Symbol: SALL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001914740.5 | XP_001914775.3 | 2982 | 60 | ||
XM_023626763.1 | XP_023482531.1 | 1663 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
