Assay Details
Gene Symbol: RTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489271.6 | XP_001489321.2 | 483 | 54 | ||
XM_023626784.1 | XP_023482552.1 | 483 | 54 | |||
XM_023626785.1 | XP_023482553.1 | 483 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
