Assay Details
Gene Symbol: ZBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604802.3 | XP_005604859.1 | 595 | 65 | ||
XM_014735300.2 | XP_014590786.1 | 2318 | 65 | |||
XM_014735301.2 | XP_014590787.1 | 2322 | 65 | |||
XM_023626795.1 | XP_023482563.1 | 2316 | 65 | |||
XM_023626796.1 | XP_023482564.1 | 2321 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
