Assay Details
Gene Symbol: GID8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493585.6 | XP_001493635.1 | 619 | 64 | ||
XM_023626896.1 | XP_023482664.1 | 398 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
