Assay Details
Gene Symbol: EEF1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626927.1 | XP_023482695.1 | 851 | 60 | ||
XM_023626928.1 | XP_023482696.1 | 722 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
