Assay Details
Gene Symbol: DNAJC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735340.2 | XP_014590826.1 | 423 | 57 | ||
XM_023626977.1 | XP_023482745.1 | 544 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
