Assay Details
Gene Symbol: PCMTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495654.5 | XP_001495704.2 | 1001 | 64 | ||
XM_023626999.1 | XP_023482767.1 | 446 | 64 | |||
XM_023627000.1 | XP_023482768.1 | 470 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
