Assay Details
Gene Symbol: PRXL2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627068.1 | XP_023482836.1 | 381 | 85 | ||
XM_023627069.1 | XP_023482837.1 | 384 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
